Merck
  • Using [1-(13) C]lactic acid for hyperpolarized (13) C MR cardiac studies.
Using [1-(13) C]lactic acid for hyperpolarized (13) C MR cardiac studies.
Magnetic resonance in medicine (2014-07-22)
Albert P Chen, Justin Y C Lau, Rohan D A Alvares, Charles H Cunningham
ABSTRACT
Hyperpolarized [1-(13) C]lactate in solution may be a clinically relevant and safe substrate for real time MR investigations of key metabolic pathways. The potential of using hyperpolarized [1-(13) C]lactate for magnetic resonance studies of cardiac metabolism in vivo was explored. Neat [1-(13) C]lactic acid was hyperpolarized using the dynamic nuclear polarization process. Cardiac MR spectroscopy experiments were performed in vivo using hyperpolarized [1-(13) C]lactate and [1-(13) C]pyruvate in solutions. A high degree of polarization was achieved for [1-(13) C]lactate in solution (16.7%). (13) C-bicarbonate was observed in rat hearts in vivo after either hyperpolarized [1-(13) C]lactate or hyperpolarized [1-(13) C]pyruvate was infused, but lower (13) C-bicarbonate to substrate ratio was observed with hyperpolarized [1-(13) C]lactate infusions. The response of (13) C-bicarbonate signal as a function of hyperpolarized [1-(13) C]lactate doses was also investigated and a saturation of (13) C-bicarbonate signal was observed at the highest dose of [1-(13) C]lactate used (0.69 mmol/kg). This study demonstrated that the use of neat [1-(13) C]lactic acid as the DNP sample is a potential alternative to [1-(13) C]pyruvic acid for cardiac hyperpolarized (13) C MR studies. Hyperpolarized [1-(13) C]lactate may enable noninvasive assessment of cardiac PDH flux in cardiac patients in the near future.
Sigma-Aldrich
L-Lactic acid-1-13C, ≥99 atom % 13C, ≥98% (CP), ≥98% (Chiral Purity, HPLC)
Sigma-Aldrich
Pyruvic-1-13C acid, ≥99 atom % 13C, ≥99% (CP)
Sigma-Aldrich
Pyruvic-1-13C acid, API for Clinical Studies, ≥99 atom % 13C