Merck
Gd2O3 nanoparticles: size-dependent nuclear magnetic resonance.
Contrast media & molecular imaging (2012-10-31)
A T M Anishur Rahman, Peter Majewski, Krasimir Vasilev
ABSTRACT
In this communication, we demonstrate that there is an optimum gadolinium oxide (Gd(2)O(3)) nanoparticle size of 2.3 nm; in the presence of Gd(2)O(3) particles smaller and larger than this critical size, the spin-lattice relaxation rate (T(1) = 1/r(1)) of water protons at 7.0 T drastically decreases. Since r(1) is directly related to the quality of magnetic resonance imaging, the results presented here have significant implications for clinical diagnostics.
Sigma-Aldrich
Gadolinium(III) oxide, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Gadolinium(III) oxide, ≥99.9%
Sigma-Aldrich
Gadolinium(III) oxide, nanopowder, <100 nm particle size (BET), 99.8% trace metals basis
Sigma-Aldrich
Gadolinium(III) oxide, powder, 99.9% trace metals basis