Merck
New polymorphs of isonicotinamide and nicotinamide.
Chemical communications (Cambridge, England) (2010-11-23)
Jinjing Li, Susan A Bourne, Mino R Caira
ABSTRACT
For each of the well-known co-crystal formers, isonicotinamide and nicotinamide, a new polymorph, obtained during attempted co-crystallisation experiments, has been fully characterized and its stability relationship with previously reported forms established.
Sigma-Aldrich
Isonicotinamide, ReagentPlus®, 99%