Merck
  • Upper electron-excited states in bioluminescence: experimental indication.
Upper electron-excited states in bioluminescence: experimental indication.
Luminescence : the journal of biological and chemical luminescence (2001-06-16)
N S Kudryasheva, E V Nemtseva, Y P Meshalkin, A G Sizykh
ABSTRACT
The involvement of upper electron-excited states in bacterial bioluminescence process was studied with excitation energy-accepting molecules. The fluorescent aromatic compounds, anthracene and 1.4-bis(5-phenyloxazol-2-yl)benzene, were chosen. Energies of their lowest excited singlet states are higher than the energy of the analogous state of the bioluminescence emitter; their absorption spectra and bioluminescence do not overlap. Hence, the excitation of these molecules by singlet-singlet energy transfer or by light absorption is excluded. Sensitized fluorescence of these compounds in the bioluminescence systems has been recorded, indicating the activity of upper electron-excited states in the bioluminescent process.
Sigma-Aldrich
1,4-Bis(5-phenyl-2-oxazolyl)benzene, suitable for scintillation, 98%
Sigma-Aldrich
1,4-Bis(5-phenyl-2-oxazolyl)benzene, BioReagent, suitable for scintillation