Merck
  • Gene expression of endoplasmic reticulum resident selenoproteins correlates with apoptosis in various muscles of se-deficient chicks.
Gene expression of endoplasmic reticulum resident selenoproteins correlates with apoptosis in various muscles of se-deficient chicks.
The Journal of nutrition (2013-03-22)
Hai-Dong Yao, Qiong Wu, Zi-Wei Zhang, Jiu-Li Zhang, Shu Li, Jia-Qiang Huang, Fa-Zheng Ren, Shi-Wen Xu, Xiao-Long Wang, Xin Gen Lei
ABSTRACT
Dietary selenium (Se) deficiency causes muscular dystrophy in various species, but the molecular mechanism remains unclear. Our objectives were to investigate: 1) if dietary Se deficiency induced different amounts of oxidative stress, lipid peroxidation, and cell apoptosis in 3 skeletal muscles; and 2) if the distribution and expression of 4 endoplasmic reticulum (ER) resident selenoprotein genes (Sepn1, Selk, Sels, and Selt) were related to oxidative damages in these muscles. Two groups of day-old layer chicks (n = 60/group) were fed a corn-soy basal diet (33 μg Se/kg; produced in the Se-deficient area of Heilongjiang, China) or the diet supplemented with Se (as sodium selenite) at 0.15 mg/kg for 55 d. Dietary Se deficiency resulted in accelerated (P < 0.05) cell apoptosis that was associated with decreased glutathione peroxidase activity and elevated lipid peroxidation in these muscles. All these responses were stronger in the pectoral muscle than in the thigh and wing muscles (P < 0.05). Relative distribution of the 4 ER resident selenoprotein gene mRNA amounts and their responses to dietary Se deficiency were consistent with the resultant oxidative stress and cell apoptosis in the 3 muscles. Expression of Sepn1, Sels, and Selt in these muscles was correlated with (r > 0.72; P < 0.05) that of Sepsecs encoding a key enzyme for biosynthesis of selenocysteine (selenocysteinyl-tRNA synthase). In conclusion, the pectoral muscle demonstrated unique expression patterns of the ER resident selenoprotein genes and GPx activity, along with elevated susceptibility to oxidative cell death, compared with the other skeletal muscles. These features might help explain why it is a primary target of Se deficiency diseases in chicks.
Sigma-Aldrich
Glutathione Peroxidase from bovine erythrocytes, lyophilized powder, ≥300 units/mg protein
Sigma-Aldrich
Glutathione Peroxidase from human erythrocytes, lyophilized powder, ≥30 units/mg protein