Merck
  • Whole-body vibration induces pain and lumbar spinal inflammation responses in the rat that vary with the vibration profile.
Whole-body vibration induces pain and lumbar spinal inflammation responses in the rat that vary with the vibration profile.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2016-08-30)
Martha E Zeeman, Sonia Kartha, Beth A Winkelstein
ABSTRACT
Whole-body vibration (WBV) is linked epidemiologically to neck and back pain in humans, and to forepaw mechanical allodynia and cervical neuroinflammation in a rodent model of WBV, but the response of the low back and lumbar spine to WBV is unknown. A rat model of WBV was used to determine the effect of different WBV exposures on hind paw behavioral sensitivity and neuroinflammation in the lumbar spinal cord. Rats were exposed to 30 min of WBV at either 8 or 15 Hz on days 0 and 7, with the lumbar spinal cord assayed using immunohistochemistry at day 14. Behavioral sensitivity was measured using mechanical stimulation of the hind paws to determine the onset, persistence, and/or recovery of allodynia. Both WBV exposures induce mechanical allodynia 1 day following WBV, but only the 8 Hz WBV induces a sustained decrease in the withdrawal threshold through day 14. Similarly, increased activation of microglia, macrophages, and astrocytes in the superficial dorsal horn of the lumbar spinal cord is only evident after the painful 8 Hz WBV. Moreover, extracellular signal-regulated kinase (ERK)-phosphorylation is most robust in neurons and astrocytes of the dorsal horn, with the most ERK phosphorylation occurring in the 8 Hz group. These findings indicate that a WBV exposure that induces persistent pain also induces a host of neuroimmune cellular activation responses that are also sustained. This work indicates there is an injury-dependent response that is based on the vibration parameters, providing a potentially useful platform for studying mechanisms of painful spinal injuries. © 2016 Orthopaedic Research Society. Published by Wiley Periodicals, Inc. J Orthop Res 34:1439-1446, 2016.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, Alexa Fluor 555 Conjugate, clone A60, from mouse, ALEXA FLUOR 555