Merck
  • Sphingomonas palmae sp. nov. and Sphingomonas gellani sp. nov., endophytically associated phyllosphere bacteria isolated from economically important crop plants.
Sphingomonas palmae sp. nov. and Sphingomonas gellani sp. nov., endophytically associated phyllosphere bacteria isolated from economically important crop plants.
Antonie van Leeuwenhoek (2020-09-20)
Munusamy Madhaiyan, Venkatakrishnan Sivaraj Saravanan, Joseph S Wirth, Tan Hian Hwee Alex, Soo-Jin Kim, Hang-Yeon Weon, Soon-Wo Kwon, William B Whitman, Lianghui Ji
ABSTRACT
In this study, two endophytic bacterial strains designated JS21-1T and S6-262T isolated from leaves of Elaeis guineensis and stem tissues of Jatropha curcas respectively, were subjected for polyphasic taxonomic approach. On R2A medium, colonies of strains JS21-1T and S6-262T are orange and yellow, respectively. Phylogenetic analyses using 16S rRNA gene sequencing and whole-genome sequences placed the strains in distinct clades but within the genus Sphingomonas. The DNA G + C content of JS21-1T and S6-262T were 67.31 and 66.95%, respectively. Furthermore, the average nucleotide identity and digital DNA-DNA hybridization values of strains JS21-1T and S6-262T with phylogenetically related Sphingomonas species were lower than 95% and 70% respectively. The chemotaxonomic studies indicated that the major cellular fatty acids of the strain JS21-1T were summed feature 8 (C18:1 ω7c and/or C18:1 ω6c), C16:0, and C14:0 2OH; strain S6-262T possessed summed feature 3 (C16:1 ω7c and/or iso-C15:0 2-OH) and summed feature 8 (C18:1 ω6c and/or C18:1 ω7c). The major quinone was Q10, and the unique polyamine observed was homospermidine. The polar lipid profile comprised of mixture of sphingoglycolipid, phosphatidylethanolamine, phosphatidylglycerol, diphosphatidylglycerol and certain uncharacterised phospholipids and lipids. Based on this polyphasic evidence, strains JS21-1T and S6-262T represent two novel species of the genus Sphingomonas, for which the names Sphingomonas palmae sp. nov. and Sphingomonas gellani sp. nov. are proposed, respectively. The type strain of Sphingomonas palmae sp. nov. is JS21-1T (= DSM 27348T = KACC 17591T) and the type strain of Sphingomonas gellani sp. nov. is S6-262T (= DSM 27346T =  KACC 17594T).
Sigma-Aldrich
Phytatray II, L × W × H 11.4 cm × 8.6 cm × 10.2 cm  in., PETG, sterile; γ-irradiated
Sigma-Aldrich
Golgicide A, ≥98% (HPLC)