Merck
  • Involvement of nitric oxide pathway in the anti-inflammatory effect of modafinil on indomethacin-, stress-, and ethanol -induced gastric mucosal injury in rat.
Involvement of nitric oxide pathway in the anti-inflammatory effect of modafinil on indomethacin-, stress-, and ethanol -induced gastric mucosal injury in rat.
European journal of pharmacology (2020-09-21)
Pegah Dejban, Faezeh Eslami, Nastaran Rahimi, Nasrin Takzare, Mohamadmostafa Jahansouz, Ahmad Reza Dehpour
ABSTRACT
Gastric ulcer is a prevalent disease with various etiologies, including non-steroidal anti-inflammatory drugs (NSAIDs), stress conditions, and alcohol, resulting in an inflammatory condition in the gastric mucosa. The aim of this study was to explore the protective effects of modafinil on gastric erosions induced by indomethacin, water-immersion stress, and alcohol in rats and to evaluate the role of nitric oxide (NO) pathway. Animals were allocated to the three experimental models of gastric ulcer - indomethacin (30 mg/kg PO), water-immersion stress, and ethanol (5 ml/kg PO). Induction of gastric ulcer in all models caused an increase in J-score (macroscopic assessment), biochemical markers, including tumor necrosis factor-alpha (TNF-α), interleukin-1beta (IL-1β), and myeloperoxidase (MPO), and microscopic destructions. Administration of modafinil (50 and 100 mg/kg i. p) significantly improved J-score in the indomethacin (P < 0.05) and stress models (P < 0.001). Moreover, the level of TNF-α IL-1β, and MPO was deceased after modafinil administration (P < 0.001). However, modafinil did not have any effects on gastric injury induced by ethanol. In addition, co-administration of L-NAME (a non-specific NO synthase inhibitor) and aminoguanidine (an inducible NO synthase inhibitor) with modafinil significantly neutralized the gastroprotective effect of modafinil in the indomethacin and water-immersion stress groups (P < 0.05, and P < 0.01; respectively), while 7-nitroindazole (a neuronal NO synthase inhibitor) did not show such reversing effects. In conclusion, modafinil possesses gastroprotective effects on the gastric lesions induced by indomethacin and stress, which are probably mediated via the inflammation inhibition and NO pathway modulation.
Sigma-Aldrich
Rat IL-1 β ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant