Merck
Leishmania parasite detection and quantification using PCR-ELISA.
Nature protocols (2010-06-12)
Tetyana Kobets, Jana Badalová, Igor Grekov, Helena Havelková, Milena Svobodová, Marie Lipoldová
ABSTRACT
This protocol describes an improved and optimized PCR-ELISA method for detection and quantification of Leishmania parasites in host tissues. Unlike other DNA-based assays, this method uses digoxigenin- and biotin-labeled primers. This eliminates the need for a separate step of hybridization of the PCR product with labeled probes. The PCR product is detected using sandwich ELISA with antidigoxigenin-detecting antibodies. Primers are complementary to the kinetoplast minicircle conserved region of parasite DNA, allowing the detection of several Leishmania species. For measurement of a wide range of parasite concentrations, +/-25 cycles were optimal. The sensitivity of this technique is 0.3 fg of parasite DNA per reaction in 40-cycle PCR-ELISA, corresponding to 0.004 parasites. DNA preparation by a standard TRI reagent procedure takes about 4 h. When DNA is prepared, a single person can test a large number of samples (at least 150) in a maximum of 7 h. This method might also be suitable for detecting and quantifying other pathogens, especially for detecting small differences in pathogen numbers.
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Deoxynucleotide Set, 100 mM, Individual dNTPs for routine PCR; 1 mL each