Merck
  • Methyljasmonate displays in vitro and in vivo activity against multiple myeloma cells.
Methyljasmonate displays in vitro and in vivo activity against multiple myeloma cells.
British journal of haematology (2012-09-14)
Steffen Klippel, Jana Jakubikova, Jake Delmore, Melissa Ooi, Douglas McMillin, Efstathios Kastritis, Jacob Laubach, Paul G Richardson, Kenneth C Anderson, Constantine S Mitsiades
ABSTRACT
Jasmonates, plant stress hormones, have been demonstrated to be effective in killing various types of cancer cells. We therefore tested if methyljasmonate (MJ) has activity against multiple myeloma (MM) in vitro and in vivo. MM cell lines and primary MM tumour cells responded to MJ in vitro at concentrations that did not significantly affect normal haematopoietic cells, without stroma-mediated resistance. Brief MJ exposures of MM cells caused release of Hexokinase 2 (HK2) from mitochondria, rapid ATP depletion, perturbation of major intracellular signalling pathways, and ensuing mainly apoptotic cell death. Sensitivity to MJ correlated with lower cellular glucose consumption and lactate production, as well as lower intracellular protein levels of HK2, phosphorylated Voltage-dependent anion channel 2/3 (pVDAC2/3) and Aldo-keto reductase family 1 member C1 (AKR1C1), which represent potential biomarkers of responsiveness to MJ treatment, especially as AKR1C1 transcript levels also correlate with clinical outcome in bortezomib- or dexamethasone-treated MM patients. Interestingly, MJ synergized with bortezomib in vitro and prolonged survival of immunocompromised mice harbouring diffuse lesions of MM.1S cells compared to vehicle-treated mice (P = 0·0046). These studies indicate that jasmonates represent a new, promising strategy to treat MM.
Sigma-Aldrich
Anti-p53 Antibody, clone BP53-12, clone BP53-12, Upstate®, from mouse