
Tiotropium for system suitability 1

Synonym(s):
Tiotropium bromide monohydrate
Empirical Formula (Hill Notation):
C19H22BrNO4S2 · H2O
CAS Number:
Molecular Weight:
490.43
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing