
Promethazine sulfoxide

Synonym(s):
(2RS)-N,N-Dimethyl-1-(10H-phenothiazin-10-yl)propan-2-amine S-oxide, Promethazine 5-sulfoxide
Empirical Formula (Hill Notation):
C17H20N2OS
CAS Number:
Molecular Weight:
300.42
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing