Poloxamer 407
Synonym(s):
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol)
CAS Number:
Products
Product Category
Brand
Manufacturer
pH Value
Physical Form
Format
Chemical Composition
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing