Skip to Content
Merck
Platinum(II) acetylacetonate ≥99.98% trace metals basis

Platinum(II) acetylacetonate

Platinum(II) acetylacetonate ≥99.98% trace metals basis
Synonym(s):
2,4-Pentanedione platinum(II) derivative, Pt(acac)2
Linear Formula:
Pt(C5H7O2)2
CAS Number:
Molecular Weight:
393.29
EC Number:

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing