
Perfluorotridecanoic acid

Linear Formula:
F3C(CF2)11CO2H
CAS Number:
Molecular Weight:
664.11
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing