Skip to Content
Merck
N,N-Diethylethylenediamine ≥99%

N,N-Diethylethylenediamine

N,N-Diethylethylenediamine ≥99%
Synonym(s):
2-Diethylaminoethylamine
Linear Formula:
(C2H5)2NCH2CH2NH2
CAS Number:
Molecular Weight:
116.20
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing