
Nitrate Reagent B

Synonym(s):
Sulfanilic acid solution, 4-Aminobenzenesulfonic acid
CAS Number:
Products
Product Category
Brand
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing