Skip to Content
Merck
Muscimol phyproof® Reference Substance

Muscimol

Muscimol phyproof® Reference Substance
Synonym(s):
3-Hydroxy-5-aminomethyl-isoxazole, 5-Aminomethyl-3-hydroxy-isoxazole, 5-Aminomethyl-3-isoxazolol, Agarine, Pantherine
Empirical Formula (Hill Notation):
C4H6N2O2
CAS Number:
Molecular Weight:
114.10
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing