
Methotrimeprazine

Synonym(s):
(−)-10-(3-[Dimethylamino]-2-methylpropyl)-2-methoxyphenothiazine, Levomeprazine
Empirical Formula (Hill Notation):
C19H24N2OS
CAS Number:
Molecular Weight:
328.47
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing