Skip to Content
Merck
L-Glutamic acid-13C5 98 atom % 13C, 95% (CP)

L-Glutamic acid-13C5

L-Glutamic acid-13C5 98 atom % 13C, 95% (CP)
Synonym(s):
(S)-2-Aminopentanedioic acid-13C5, L-Glutamic acid-UL-13C, 13C Labeled L-glutamic acid, Glu-13C5
Linear Formula:
HO213C(13CH2)213CH(NH2)13CO2H
CAS Number:
Molecular Weight:
152.09

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing