
Kaempferitrin

Synonym(s):
3,7-Bis((6-deoxy-α-L-mannopyranosyl)oxy)-5-hydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Kaempferol 3,7-dirhamnoside, Lespedin, Lespenefril, Lespenephryl
Empirical Formula (Hill Notation):
C27H30O14
CAS Number:
Molecular Weight:
578.52
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing