![Formaldehyde solution about 37% GR for analysis stabilized with about 10% methanol ACS,Reag. Ph Eur](/deepweb/assets/sigmaaldrich/product/structures/217/258/f662268b-9b76-487e-b611-591a8b1eaca1/640/f662268b-9b76-487e-b611-591a8b1eaca1.png)
Formaldehyde solution about 37%
![Formaldehyde solution about 37% GR for analysis stabilized with about 10% methanol ACS,Reag. Ph Eur](/deepweb/assets/sigmaaldrich/product/structures/217/258/f662268b-9b76-487e-b611-591a8b1eaca1/640/f662268b-9b76-487e-b611-591a8b1eaca1.png)
Synonym(s):
Formaldehyde solution, Formalin
Linear Formula:
HCHO
CAS Number:
Molecular Weight:
30.03
Products
Formula Weight
Physical Form
Agency
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing