
Econazole Related Compound C

Synonym(s):
1-(4-Chlorobenzyl)-3-[(2RS)-2-[(4-chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethyl]imidazolium chloride
Empirical Formula (Hill Notation):
C25H21Cl5N2O
Molecular Weight:
542.71
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2