
(+)-Isocorydine hydrochloride

Synonym(s):
11-Hydroxy-1,2,10-trimethoxyaporphine, Isocorydine hydrochloride, Luteanine
Empirical Formula (Hill Notation):
C20H23NO4 · HCl
CAS Number:
Molecular Weight:
377.86
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing