![Dimenhydrinate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/295/942/bcc1b8c2-fdc9-4719-b675-7cc3a60941ce/640/bcc1b8c2-fdc9-4719-b675-7cc3a60941ce.png)
Dimenhydrinate
![Dimenhydrinate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/295/942/bcc1b8c2-fdc9-4719-b675-7cc3a60941ce/640/bcc1b8c2-fdc9-4719-b675-7cc3a60941ce.png)
Synonym(s):
N-(2-Diphenylmethoxyethyl)-N,N-dimethylammonium 8-chlorotheophyllinate, Dimenhydrinate
Empirical Formula (Hill Notation):
C17H21NO · C7H7ClN4O2
CAS Number:
Molecular Weight:
469.96
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4