Skip to Content
Merck
Diethyl maleate ≥96%, FG

Diethyl maleate

Diethyl maleate ≥96%, FG
Synonym(s):
Maleic acid diethyl ester
Linear Formula:
C2H5OCOCH=CHCOOC2H5
CAS Number:
Molecular Weight:
172.18
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing