Chymostatin
Synonym(s):
N-(Nα-Carbonyl-Cpd-X-Phe-al)-Phe (Cpd = capreomycidine) (capreomycidine = [S,S]-α-(2-Iminohexahydro-4-pyrimidyl)glycine), Chymostatin
CAS Number:
Products
Brand
Biological Source
Manufacturer
Physical Form
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing