
Cereulide solution

Empirical Formula (Hill Notation):
C57H96N6O18
CAS Number:
Molecular Weight:
1153.40
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing