
(−)-Carvone

Synonym(s):
(−)-p-Mentha 6,8-diene 2-one, (R)-5-Isopropenyl-2-methyl-2-cyclohexenone, Carvol
Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing