Skip to Content
Merck
Brivaracetam ≥98% (HPLC)

Brivaracetam

Brivaracetam ≥98% (HPLC)
Synonym(s):
(αS,4R)-α-Ethyl-2-oxo-4-propyl-1-pyrrolidineacetamide, (2S)-2-[(4R)-2-Oxo-4-propylpyrrolidin-1-yl]butanamide, (S)-2-((R)-2-Oxo-4-propylpyrrolidin-1-yl)butanamide, UCB 34714, UCB-34714, UCB34714
Empirical Formula (Hill Notation):
C11H20N2O2
CAS Number:
Molecular Weight:
212.29

Products

Brand
Color
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing