Boc-Homophe-OH
Synonym(s):
(S)-2-(Boc-amino)-4-phenylbutyric acid, Boc-L-homophenylalanine
Linear Formula:
C6H5CH2CH2CH[NHCOOC(CH3)3]COOH
CAS Number:
Molecular Weight:
279.33
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing