
Anti-SARS-CoV-2 (COVID-19) NSP13 (Helicase) antibody produced in rabbit

Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-2 of 2