
Amisulpride for system suitability

Synonym(s):
Amisulpride, 4-Amino-N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-(ethylsulfonyl)-2-methoxybenzamide, DAN-2163, Deniban
Empirical Formula (Hill Notation):
C17H27N3O4S
CAS Number:
Molecular Weight:
369.48
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2