
5-Methyl-2-thiophenecarboxaldehyde

Synonym(s):
NSC 87542
Empirical Formula (Hill Notation):
C6H6OS
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing