
5′-Deoxy-5′-(methylthio)adenosine

Synonym(s):
MTA
Empirical Formula (Hill Notation):
C11H15N5O3S
CAS Number:
Molecular Weight:
297.33
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing