
5-Bromo-4-chloro-3-indolyl phosphate disodium salt

Synonym(s):
BCIP®, X-phosphate disodium salt
Linear Formula:
C8H4BrClNO4P · 2Na
CAS Number:
Molecular Weight:
370.43
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing