
3,5-Dimethyl-4H-1,2,4-triazole

Empirical Formula (Hill Notation):
C4H7N3
CAS Number:
Molecular Weight:
97.12
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing