
2-(2-Hydroxyethyl)-1-methylpyrrolidine

Synonym(s):
1-Methyl-2-pyrrolidine ethanol
Empirical Formula (Hill Notation):
C7H15NO
CAS Number:
Molecular Weight:
129.20
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing