Skip to Content
Merck
1-Heptanol analytical standard, ≥99.5% (GC)

1-Heptanol

1-Heptanol analytical standard, ≥99.5% (GC)
Synonym(s):
Heptyl alcohol
Linear Formula:
CH3(CH2)6OH
CAS Number:
Molecular Weight:
116.20
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing