Skip to Content
Merck
1-Aminocyclopropanecarboxylic acid ≥98% (TLC), powder

1-Aminocyclopropanecarboxylic acid

1-Aminocyclopropanecarboxylic acid ≥98% (TLC), powder
Synonym(s):
ACPC
Empirical Formula (Hill Notation):
C4H7NO2
CAS Number:
Molecular Weight:
101.10

Products

Product Category
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing