
1,2-Dimyristoyl-sn-glycerol

Synonym(s):
(S)-1,2-Dimyristin, (S)-Glycerol 1,2-dimyristate, 1,2-Ditetradecanoyl-sn-glycerol, DMG
Empirical Formula (Hill Notation):
C31H60O5
CAS Number:
Molecular Weight:
512.81
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing