Tropomyosin
Tropomyosin together with troponin, regulate the binding of myosin to actin. Tropomyosin is a dimeric coiled-coil protein forming continuous polymers along the myosin-binding groove of actin. Varoius tropomyosin isoforms help to confer structure and function to actins in the cytoskeleton and in striated muscle function.
![Tropomyosin Tropomyosin](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/research-and-disease-areas/cell-signaling/tropomyosin.gif)