USP HPLC Analysis of Ampicillin Sodium on Ascentis® Express C18
![USP HPLC Analysis of Ampicillin Sodium on Ascentis® Express C18 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/372/250/1224d1e0-b7fb-4a89-8219-bb1faad05c69/800/1224d1e0-b7fb-4a89-8219-bb1faad05c69.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Ascentis Express C18, 10 cm × 4.6 mm I.D., 5 μm particles (50536-U)
mobile phase
[A:B:C:D] [A] acetonitrile, [B]water, [C] 1M Potassium phosphate monobasic in water, [D] 1N acetic acid in water (80:909:10:1)
flow rate
1.0 mL/min
column temp.
30 °C
detector
UV, 254 nm
injection
10 μL
sample
1 mg/mL in mobile phase
Description
Analysis Note
This application was generated by RTC on an Ascentis Express C18.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany