Merck
LC/MS Analysis of Polar Amino Acids on Ascentis® Si (HILIC Mode)
LC/MS Analysis of Polar Amino Acids on Ascentis® Si (HILIC Mode) application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Si HPLC Column
5 μm particle size, L × I.D. 10 cm × 2.1 mm
standard
Product No.
Description
Pricing
L-Asparagine
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
L-Threonine
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Serine
European Pharmacopoeia (EP) Reference Standard
Aspartic acid
United States Pharmacopeia (USP) Reference Standard
Glutamine
United States Pharmacopeia (USP) Reference Standard
Glutamic acid
United States Pharmacopeia (USP) Reference Standard
CONDITIONS
column
Ascentis Si, 10 cm x 2.1 mm I.D., 5 μm particles (581508-U)
mobile phase
[A] 100 mM ammonium formate, pH 3.0 with formic acid; [B] water; [C] acetonitrile
gradient
5% A constant; 10 to 16% B, 85 to 79% C in 20 min
flow rate
0.3 mL/min
column temp.
35 °C
detector
ESI(+)
injection
2 μL
sample
0.1 g/L each in mobile phase
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany