Merck
HPLC Analysis of α-Methylbenzylamine Enantiomers (DANSYL Derivatives) on Astec® CYCLOBOND I 2000 DMP
HPLC Analysis of α-Methylbenzylamine Enantiomers (DANSYL Derivatives) on Astec® CYCLOBOND I 2000 DMP application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CYCLOBOND I 2000 DMP Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
standard
Product No.
Description
Pricing
(S)-(−)-α-Methylbenzylamine
for chiral derivatization, LiChropur, ≥99.0%
(R)-(+)-α-Methylbenzylamine
for chiral derivatization, LiChropur, ≥99.0%
(R)-(+)-α-Methylbenzylamine
purum, ≥98.0% (sum of enantiomers, GC)
(R)-(+)-α-Methylbenzylamine
98%
(S)-(−)-α-Methylbenzylamine
98%
α-Methylbenzylamine
99%
CONDITIONS
column
CYCLOBOND I 2000 DMP, 25 cm x 4.6 mm I.D., 5 μm particles (20724AST)
mobile phase
ethanol
flow rate
1.2 mL/min
column temp.
22 °C
detector
UV, 254 nm
injection
1 μL
sample
3 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null