Merck
HPLC Analysis of Fusion Protein on Zenix®-C SEC-300
HPLC Analysis of Fusion Protein on Zenix®-C SEC-300 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Zenix®-C SEC-300 Gel Filtration Column, 3 μm
L × I.D. 30 cm × 7.8 mm, 3 μm particle size, stainless steel
CONDITIONS
column
Zenix-C SEC-300, 30 cm x 7.8 mm I.D., 3 μm particles (Z777092)
mobile phase
150 mM sodium phosphate buffer, pH 7.0 + 200 mM NaCl
flow rate
1.0 mL/min
pressure
1102 psi (76 bar)
column
25 °C
detector
UV, 214 nm
injection
10 μL
sample
1 mg/mL fusion protein
Description
Legal Information
Zenix is a registered trademark of Sepax Technologies