HPLC Analysis of FMOC-Serine Enantiomers on Astec® (R,R) P-CAP™
![HPLC Analysis of FMOC-Serine Enantiomers on Astec® (R,R) P-CAP™ application for HPLC](/deepweb/assets/sigmaaldrich/product/images/367/915/b06aea66-31ea-4d52-8ad2-037e9f301acb/800/b06aea66-31ea-4d52-8ad2-037e9f301acb.jpg)
Materials
standard
Product No.
Description
Pricing
L-Serine
from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%CONDITIONS
column
Astec (R,R) P-CAP, 25 cm x 4.6 mm I.D., 5 μm particles (31024AST)
mobile phase
20 mM ammonium acetate in: (A) acetonitrile; (B) methanol; (70:30, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
0.5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
P-CAP is a trademark of Sigma-Aldrich Co. LLC