GC Analysis of trans C18:1 FAMEs in Partially Hydrogenated Vegetable Oil (PHVO) on SP®-2560 after HPLC Fractionation using Silver Ion
![GC Analysis of trans C18:1 FAMEs in Partially Hydrogenated Vegetable Oil (PHVO) on SP®-2560 after HPLC Fractionation using Silver Ion suitable for GC](/deepweb/assets/sigmaaldrich/product/images/259/774/552c5706-1ccc-46c1-ba4e-b51ca822b339/800/552c5706-1ccc-46c1-ba4e-b51ca822b339.jpg)
CONDITIONS
column
SP-2560, 100 m x 0.25 mm I.D., 0.20 μm (24056)
oven
180 °C isothermal
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
hydrogen, 1 mL/min
injection
1 μL, 100:1 split
liner
4 mm I.D., split liner with cup (2051001)
sample
trans fraction of partially hydrogenated vegetable oil FAMEs, prepared using Ag-ion fractionation
Description
Legal Information
SP is a registered trademark of Sigma-Aldrich Co. LLC