GC Analysis of Amphetamines in Urine on a 100% Methyl Column after SPME using 100 μm PDMS Fiber
![GC Analysis of Amphetamines in Urine on a 100% Methyl Column after SPME using 100 μm PDMS Fiber suitable for GC, application for SPME](/deepweb/assets/sigmaaldrich/product/images/333/907/5c70488d-cf53-43cb-9f2a-a8de278bb6aa/800/5c70488d-cf53-43cb-9f2a-a8de278bb6aa.jpg)
Materials
SPME fiber
Product No.
Description
Pricing
SPME fiber assembly Polydimethylsiloxane (PDMS)
df 100 μm(PDMS, needle size 24 ga, for use with manual holderCONDITIONS
sample/matrix
1 mL urine + 0.7g potassium carbonate in 20 mL headspace vial, equilibrated at 80°C, 30 min
SPME fiber
polydimethylsiloxane, 100 μm (57300-U)
derivatization
methyl bis-trifluoroacetamide (headspace, 0.5 min, ambient)
extraction
headspace, 3-5 min, 80 °C
desorption process
1 min, 270 °C
column
100% methyl polysiloxane, 12.5 m × 0.2 mm I.D., 0.33 μm (Supelco equivalent, Equity-1, 28041-U)
oven
60 °C (1 min) to 140 °C (4 min) at 30 °C/min, then to 276 °C at 20 °C/min, 4 min
inj. temp.
splitless (closed 1 min), 270 °C
detector
MS, full scan