Merck
US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm) suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.50 μm
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.50 μm (28473-U)
oven
40 °C (1 min), 12 °C/min to 250 °C, 25 °C/min to 340 °C (4.5 min)
inj. temp.
250 °C
MSD interface
340 °C
scan range
m/z = 40-450
carrier gas
helium, 1.5 mL/min constant
injection
1.0 μL, splitless (0.50 min)
liner
4 mm I.D., single taper
sample
50 ng on-column of a 62 component semivolatile standard and 8 surrogate compounds, plus 6 internal standards (at 40 ng on-column)
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany